Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Renin R Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$670.00
Specifications
Antigen | Renin R |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Renin R Polyclonal specifically detects Renin R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
Renin R | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
10159 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NSLSRNNEVDLLFLSELQVLHDISSLLSRHKHLAKDHSPDLYSLELAGLDEIGKRYGEDSEQFRDASKILVDALQKFADDMYSLYGGNAVVELVTVKSFDTSLIRKTRTILEAKQAKNPASPYNLAYKYNFEYSVVFN | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
APT6M8-9, ATP6M8-9MRXE, ATPase H(+)-transporting lysosomal accessory protein 2, ATPase H(+)-transporting lysosomal-interacting protein 2, ATPase, H+ transporting, lysosomal accessory protein 2, ATPase, H+ transporting, lysosomal interacting protein 2, CAPER, ELDF10, Embryonic liver differentiation factor 10, ER-localized type I transmembrane adaptor, H+ transporting, lysosomal (vacuolar proton pump) membrane sectorassociated protein M8-9, M8-9, MGC99577, MSTP009, N14F, renin receptor, vacuolar proton ATP synthase membrane sector associated protein M8-9, V-ATPase M8.9 subunit, XMRE | |
ATP6AP2 | |
IgG | |
Affinity Purified | |
Specificity of human Renin R antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title