Learn More
Sartorius™ ReproEasy Performance Test Pads
Shop All Sartorius Products
Click to view available options
Type:
Moisture Analyzer Test Pads, 10/Pk.
Description
Test pads for the periodic review of the performance of MA37/160 moisture analyzer.
- Allows you to quickly confirm whether a device is working properly and delivers correct results.
- Use together with the performance test function in moisture analyzer.
Specifications
Specifications
| For Use With (Equipment) | MA37/160 Moisture Analyzer |
| Quantity | 20/Pk. |
| Type | Moisture Analyzer Test Pads, 10/Pk. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov