Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RER1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP237700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237700 Supplier Novus Biologicals Supplier No. NBP237700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RER1 Polyclonal specifically detects RER1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen RER1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_008964.3
Gene Alias protein RER1, RER1 retention in endoplasmic reticulum 1 homolog (S. cerevisiae)
Gene Symbols RER1
Host Species Rabbit
Immunogen This antibody maps to a region between residue 146 to 196 of human Retention in Endoplasmic Reticulum 1 Homolog using the numbering given in entry NP_008964.3 (GeneID 11079).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11079
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.