Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative E1A DNA Fragment Length Kits

The resDNASEQ Quantitative E1A DNA Fragment Length Kit is a quantitative PCR (qPCR)-based system that measures and quantitates the fragment lengths of residual host-cell DNA by targeting the E1A gene in HEK 293 cell lines.

$4795.00 - $5400.00

Specifications

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell HEK
Label or Dye FAM
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
A51969
View Documents
Applied Biosystems™
A51969
Assay Kit only
Each for $4,795.00
Only null left
Add to Cart
 
A51970
View Documents
Applied Biosystems™
A51970
Assay Kit and Sample Preparation
Each for $5,400.00
Only null left
Add to Cart
 
Description

Description

The resDNASEQ Quantitative E1A DNA Fragment Length Kit is a quantitative PCR (qPCR)-based system that measures and quantitates the fragment lengths of residual host-cell DNA by targeting the E1A gene in HEK 293 cell lines, which are predominantly used in the development of gene therapy, cell-based vaccines, and similar biotherapeutics. Reliable and rapid, the resDNASEQ kit enables sensitive and specific quantitation of different fragment lengths of the E1A gene within the residual host-cell DNA. This performance helps ensure a high degree of confidence in the obtained quantitation data.

Features of the resDNASEQ Quantitative E1A DNA Fragment Length Kit include:
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, three TaqMan primer/probe mixes, and a single E1A plasmid DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Integrated workflow solution
The resDNASEQ Quantitative E1A DNA Fragment Length Kit is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing. The kit is compatible with the PrepSEQ Residual DNA Sample Preparation Kit, which offers both a manual and automated sample preparation workflow. The resDNASEQ kit has been validated on the Applied Biosystems 7500 Fast Real-Time PCR System and the QuantStudio 5 Real-Time PCR System. Data analysis is streamlined using AccuSEQ Real-Time PCR Detection Software that provides accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance.

Easy to use
With the resDNASEQ Quantitative E1A DNA Fragment Length Kit, you perform qPCR analysis to quantitate the copy numbers of three different fragment size ranges in your test samples and compare that against standard curves generated with known amounts of specific fragments of the E1A DNA standard. The resDNASEQ kit includes almost everything you need to carry out these steps, from master mix to TaqMan primer/probe mix to E1A DNA standards, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of fragments of three different lengths of E1A DNA, assay results must be unaffected by the size of non-specific DNA present in the test sample. With the resDNASEQ system you get consistent assay performance across the three E1A fragment sizes.

Specifications

Specifications

Primer-probe
qPCR
FAM
100 reactions
Kit
HEK
resDNASEQ™
100 Reactions
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.