Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ resDNASEQ™ Quantitative HEK293 DNA Kits

The resDNASEQ Quantitative HEK293 DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host-cell DNA from HEK293 cell lines.

$3460.00 - $3855.00

Specifications

Detection Method Primer-probe
Format Kit
PCR Method qPCR
Host Cell HEK
Label or Dye FAM
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
A46014
View Documents
Applied Biosystems™
A46014
Assay Kit only
Each for $3,460.00
Only null left
Add to Cart
 
A46565
View Documents
Applied Biosystems™
A46565
Assay Kit and Sample Preparation
Each for $3,855.00
Only null left
Add to Cart
 
Description

Description

The resDNASEQ Quantitative HEK293 DNA Kit is a quantitative PCR (qPCR)-based system for the detection of host-cell DNA from HEK293 cell lines used in the development of gene therapy, cell-based vaccines, and similar biotherapeutics. Reliable and rapid, the resDNASEQ system enables sensitive and specific quantitation of HEK293 cell DNA. This performance helps ensure a high degree of confidence in quantitation data obtained from a broad range of sample types, from in-process samples with different sample matrices to purified final product.

Features of the resDNASEQ Quantitative HEK293 DNA Kit include:
• Highly sensitive quantitation using proven TaqMan real-time qPCR technology
• Consistent performance across the expected range of DNA fragment sizes
• Includes master mix, TaqMan primer/probe mix, and HEK293 genomic DNA standard

This kit is available with or without sample preparation reagents in the form of a PrepSEQ Residual DNA Sample Preparation Kit, which is optimized for highly efficient residual DNA recovery from complex mixtures of proteins, buffers, and salts.

Integrated workflow solution
The resDNASEQ Quantitative HEK293 DNA Kit is part of an integrated workflow for impurity and contaminant testing during biopharmaceutical manufacturing. The kit is compatible with the PrepSEQ Residual DNA Sample Preparation Kit, which offers both a manual and automated sample preparation workflow. The resDNASEQ kit has been validated on the Applied Biosystems 7500 Fast Real-Time PCR System and the Applied Biosystems QuantStudio 5 Real-Time PCR System. Data analysis is streamlined using AccuSEQ Real-Time PCR Detection Software that provides accurate quantitation and security, audit, and e-signature capabilities to help enable 21 CFR Pt 11 compliance.

Easy to use
With the resDNASEQ Quantitative HEK293 DNA Kit, you perform qPCR analysis to compare the amount of HEK293 DNA in your test samples to a standard curve generated with known amounts of purified HEK293 DNA standard. The resDNASEQ kit includes almost everything you need to carry out these steps, from master mix to TaqMan primer/probe mix to HEK293 genomic DNA standard, as well as step-by-step instructions.

Consistent performance
For accurate quantitation of residual HEK293 DNA, assay results must be unaffected by the size of the DNA molecules present in the test sample. With the resDNASEQ system you get consistent assay performance across a range of DNA fragment sizes.

*Due to the high degree of similarity between genomes, DNA from closely-related primate species, such as monkey (Vero), chimpanzee, and orangutan, are expected be detected by the resDNASEQ HEK293 Quantitation Kit.

Specifications

Specifications

Primer-probe
qPCR
FAM
100 reactions
Kit
HEK
resDNASEQ™
100 Reactions
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.