Learn More
Eppendorf™ Research™ plus Mechanical Multi-Channel Pipette PROMO
Catalog No. p-200055547 Shop All Eppendorf North America Products
Click to view available options
Volume (Metric):
1 to 20 μL
5 to 100 μL
Number of Channels:
16
24
Save up to 30% on qualifying packages that include one Eppendorf™ Research™ plus Multichannel Mechanical Pipette and two packs of Eppendorf™ twin.tec™ 384-Well PCR Plates (Cat. No. E951020702).
Promo:
Save up to 30% on qualifying combination packages that include one Eppendorf™ Research™ plus Multichannel Mechanical Pipette and two packs (25 plates per pack) of Eppendorf™ twin.tec™ 384-Well PCR Plates (Cat. No. E951020702).
Choose a package that features a:
  • Research plus 16-Channel Pipette, 1 to 20 μL or 5 to 100 μL or
  • Research plus 24-Channel Pipette, 1 to 20 μL or 5 to 100 μL
NOTE: Savings based on the cost of purchasing the products separately. Discounts taken off list price.
Research™ plus Multichannel Mechanical Pipette Features
  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume disp
  • Calibration display and adjustment opening for specific liquids and volumes
  • Has ability to be autoclaved without disassembling
twin.tec™ 384-Well PCR Plates (Cat. No. E951020702) Features
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
TRUSTED_SUSTAINABILITY
Specifications
Battery Type None
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Light Pink
Number of Channels 16
Product Line Research Plus
Type Mechanical Pipette
Volume (Metric) 1 to 20 μL
Display 4 Digits
Finger Style Ergonomic
Includes Box 2.0 with 384 pipette tips and 2 free boxes 384-well, clear, Eppendorf twin.tec™ skirted PCR plates (25 plates per box)
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.