Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resistin Antibody (HRES106), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB120686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120686 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120686 Supplier Novus Biologicals Supplier No. NBP311678AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Resistin Monoclonal specifically detects Resistin in Human samples. It is validated for ELISA, Direct ELISA.

Specifications

Antigen Resistin
Applications ELISA
Classification Monoclonal
Clone HRES106
Conjugate Alexa Fluor 488
Dilution ELISA, Direct ELISA
Formulation 50mM Sodium Borate
Gene Alias Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1
Host Species Mouse
Immunogen Recombinant human resistin.
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 56729
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.