Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus A2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB344920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344920 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344920 Supplier Novus Biologicals Supplier No. NB10093588F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Respiratory Syncytial Virus A2 Polyclonal antibody specifically detects Respiratory Syncytial Virus A2 in Human, Virus samples. It is validated for Western Blot

Specifications

Antigen Respiratory Syncytial Virus A2
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV
Host Species Rabbit
Immunogen Reacts with residues 180-198 TCWAICKRIPNKKPGKK of the human RSV G protein A2 strain.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.