Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Antibody (8B10cc)
SDP

Catalog No. NB12010016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12010016 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12010016 Supplier Novus Biologicals Supplier No. NB12010016

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Monoclonal specifically detects Respiratory Syncytial Virus in Virus samples. It is validated for ELISA, Lateral Flow Assay.

Specifications

Antigen Respiratory Syncytial Virus
Applications ELISA
Classification Monoclonal
Clone 8B10cc
Conjugate Unconjugated
Dilution ELISA 1:2000
Gene Alias Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified RSV, strain Long.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Nucleoprotein (N) protein specific. NB120-10016 reacts only with mature virions with completely assembled nucleocapsids. It fails to reorganize the N-protein epitope of immature and destroyed virions, which indicates conformation dependence of the NB120-10016 binding site.
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.