Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Antibody (9C5cc), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB359771 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359771 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359771 Supplier Novus Biologicals Supplier No. NB1107545AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Monoclonal antibody specifically detects Respiratory Syncytial Virus in Virus samples. It is validated for ELISA, Lateral Flow

Specifications

Antigen Respiratory Syncytial Virus
Applications ELISA, Lateral Flow
Classification Monoclonal
Clone 9C5cc
Conjugate Alexa Fluor 594
Dilution ELISA, Lateral Flow Assay
Formulation 50mM Sodium Borate
Gene Alias Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified RSV, strain Long.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.