Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus fusion protein Antibody (10B6), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10063020U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063020U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063020U Supplier Novus Biologicals Supplier No. NB10063020UV

Mouse Monoclonal Antibody

Respiratory Syncytial Virus fusion protein Monoclonal specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen Respiratory Syncytial Virus fusion protein
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 10B6
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA
Gene Alias RSV fusion protein
Host Species Mouse
Immunogen Purified Respiratory Syncytial Virus (RSV)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognises the Respiratory Syncytial Virus (RSV) Fusion protein, also known as F-protein. The F-protein is a membrane-associated protein found in the membrane of infected respiratory epithelial cells.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.