Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus fusion protein Antibody (RSV3216 (B016)), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB248764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB248764 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB248764 Supplier Novus Biologicals Supplier No. NB10064495AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus fusion protein Monoclonal antibody specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Respiratory Syncytial Virus fusion protein
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone RSV3216 (B016)
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias RSV fusion protein
Host Species Mouse
Immunogen Bovine Respiratory Syncytial Virus fusion protein strains: 127, SNK and 9007. Human RSV strains: Long, Randall, 8/60, and A/2.
Purification Method Protein A purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.