Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Glycoprotein F Antibody (11-2-F3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250412AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250412AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250412AH Supplier Novus Biologicals Supplier No. NBP250412AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Glycoprotein F Monoclonal specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Respiratory Syncytial Virus Glycoprotein F
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 11-2-F3
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This antibody reacts with the RS virus fusion glycoprotein F (also named VP70 or F0 protein), specifically to a subunit of the F protein, F1 (also named VGP48 or 48kDa), from both subgroups A and B of RS virus.This antibody has a high neutralising activity; 90% plaque neutralization titre of 102.2 (Gimenez et al 1996). This property makes it potentially useful for the preparation of reagents for prevention and/or therapeutic use (for example, vaccines or new-generation of man-made antibodies). The characterisation of the epitope of this antibody which induce high neutralising activity will be useful for the preparation of reagents (for example vaccine) to prevent RS virus infection. A RS virus vaccine is not yet available. A vaccine could be used to increase the maternal immunity to RS virus, which would then be passed to the newborn baby to provide passive immunity.The further characterisation of the epitope recognised by this antibody, which possesses a high neutralising activity, will be useful for the preparation of reagents to prevent RS virus infection. New-generation man-made antibodies, based on the 11-2-F3 antibody, could complement the effectiveness of polivizumab (Synagis), which is used to prevent but not treat established RS virus infections (Nature Biotechnology, 2013, 31, 952).
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.