Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Glycoprotein F Antibody (11-5-G9), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309474 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309474 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309474 Supplier Novus Biologicals Supplier No. NBP250393MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Functional Assay

Specifications

Antigen Respiratory Syncytial Virus Glycoprotein F
Applications Western Blot, ELISA, Immunocytochemistry, Functional Assay
Classification Monoclonal
Clone 11-5-G9
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Functional
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.