Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Glycoprotein G Antibody (133), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20411A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20411A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20411A32 Supplier Novus Biologicals Supplier No. NBP250411AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Glycoprotein G Monoclonal specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Respiratory Syncytial Virus Glycoprotein G
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 133
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Human RSV strain A2 infected HeLa cells
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.