Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Glycoprotein G Rabbit anti-Virus, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB182657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB182657 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB182657 Supplier Novus Biologicals Supplier No. NBP306595AF594
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Respiratory Syncytial Virus Glycoprotein G Polyclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen Respiratory Syncytial Virus Glycoprotein G
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Respiratory Syncytial Virus Glycoprotein G (His 67-Arg 297; AAB59857.1).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.