Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus M2-1 Protein Antibody (5H5), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250481AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250481AH 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250481AH Supplier Novus Biologicals Supplier No. NBP250481AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus M2-1 Protein Monoclonal antibody specifically detects Respiratory Syncytial Virus M2-1 Protein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen Respiratory Syncytial Virus M2-1 Protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 5H5
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV
Host Species Mouse
Immunogen hRSV strain A2 infected HeLa cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This antibody has broad specificity for hRSV of both sub-groups A and B.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.