Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus M2-1 Protein Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB212903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212903 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212903 Supplier Novus Biologicals Supplier No. NB10093589AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Respiratory Syncytial Virus M2-1 Protein Polyclonal antibody specifically detects Respiratory Syncytial Virus M2-1 Protein in Virus samples. It is validated for Western Blot

Specifications

Antigen Respiratory Syncytial Virus M2-1 Protein
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Fusion glycoprotein F0, Fusion glycoprotein F1, Fusion glycoprotein F2, RS virus, RSV
Host Species Rabbit
Immunogen Reacts with residues 81-95 [LESYIGSINNITKQSA] of the human Respiratory Syncytial Virus M2 protein, which is a CTL epitope. 93% identity to bovine RSV M2.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.