Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Nucleoprotein Antibody (1C3), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309267 Supplier Novus Biologicals Supplier No. NBP250534MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Nucleoprotein Monoclonal antibody specifically detects Respiratory Syncytial Virus Nucleoprotein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Respiratory Syncytial Virus Nucleoprotein
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1C3
Conjugate mFluor Violet 500 SE
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen hRSV strain A2 infected HeLa cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.