Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Subgroup A G Glycoprotein Antibody (1C2), HRP, Novus Biologicals™
SDP

Catalog No. NBP250310H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250310H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250310H Supplier Novus Biologicals Supplier No. NBP250310H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Respiratory Syncytial Virus Subgroup A G Glycoprotein Monoclonal specifically detects Respiratory Syncytial Virus Subgroup A G Glycoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Respiratory Syncytial Virus Subgroup A G Glycoprotein
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1C2
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with No Preservative
Host Species Mouse
Immunogen hRSV strain A2 infected HeLa cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.