Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Rest Stops™ Enrichment Device, Certified
SDP

Catalog No. p-8401020
Encompass
Click to view available options
Size:
Small
Length (English):
3 in.
Width (English):
3 in.

Provides a comfortable resting area

  • Offers paw relief from wire bottom floors
  • Useful to place special needs diets or enrichment treats at cage level
  • Made of transparent high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)

Specifications

Autoclavable Autoclavable
Product Type Rest Stop
Color Smoke Gray
Material Polycarbonate
For Use With (Application) Useful to place special needs diets or enrichment treats at cage level
Size Small
Length (English) 3 in.
Width (English) 3 in.
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.