Description
- Available for gas, oxygen, and stainless steel services
- Roll Dimensions: 260 x 1/2 in.
- Maximum temperature: 260°C
Specifications
Specifications
| Type | Tape |
| Material | PTFE |
| Description | Color: Green |
| For Use With (Application) | Oxygen Service |
| Operating Temperature (Metric) | Max: 260°C |
| Quantity | 1 Roll |