Learn More
Restek™ 20 mm Pressure Release Aluminum Seals with Septa, Assembled
Shop All Restek Corporation Products
Click to view available options
Septum:
PTFE, Butyl Rubber
PTFE, Silicone
No. per Pack:
100
1000
Description
- Color: Silver
- Material: PTFE/Gray Butyl Rubber Septa
Specifications
Specifications
| Color | Silver |
| Material | Aluminum |
| Septum | PTFE, Silicone |
| No. per Pack | 100 |
| Type | Crimp Cap |
| Closure Size | 20 mm |