Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Accuform Micro-Vials with Screw Threads
SDP

Catalog No. p-3180980 Shop All Restek Corporation Products
Click to view available options
Volume (Metric):
1 mL
3 mL
Closure Size:
13-425
20-425

Catalog No. 06-718-386

Supplier: Restek™ 21051

Work with Mininert™ sampling valves

  • Tapered for high recovery of contents
  • With graduated marking spot
  • Material: Borosilicate glass

Specifications

Material Borosilicate Glass
Volume (Metric) 3 mL
For Use With (Application) Mininert™ sampling valves
Graduated Yes
Base Style Tapered
Closure Type Screw Cap (Open-Top)
Quantity 12 Pk.
Closure Size 20-425
Marking Spot Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.