Learn More
Restek™ ASI Check Valves for Agilent 1050 and 1100
SDP
Catalog No. 06712730 Shop All Restek Corporation Products
Click to view available options
Type:
Outlet Cartridge
Outlet Check Valve (Cartridge and Housing)
Outlet Housing
Material:
Ceramic Ball and Seat
N/A
Sapphire and Ruby Ball and Seat
For Agilent 1050, 1100 HPLC Systems.
  • Valves do not leak even at pressures up to 12,000psi (82,737 kPa)
  • Valves close properly even without a high backpressure force
  • Cartridge design allows easy, convenient replacement of worn or damaged valves
Specifications
For Use With (Equipment) Agilent 1050 and 1100 HPLC System
Material Ceramic Ball and Seat
Type Outlet Cartridge
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.