Learn More
Restek™ ASI Piston Seals
Shop All Restek Corporation Products
Click to view available options
Type:
ASI Piston Seal
Piston Seal
Description
- Constructed of PTFE material with an additive that reduces seal's tendency to “cold flow” and improves their wear characteristics
- More resistant to any HPLC solvent than UHMWPE seals
Specifications
Specifications
For Use With | Agilent 1050 and 1100 HPLC system |
Material | PTFE |
Type | ASI Piston Seal |