Learn More
Description
- Low bleed levels
- Longer column lifetimes
- Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
- Bonded packings usually stabilize in 30 minutes or less
- Detectors can be operated at higher sensitivities due to low-bleed
- Reduces need for data system bleed compensation
- More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
- No sagging due to thorough cross-linking of material
- Mesh: 100/120 Silcoport W
Order Info
Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.
Column Configuration Key: General - General Configuration; Agilent - Agilent 5880, 5890, 5987, 6890, 7890; PE Auto Sys - PE Auto System 8300, 8400, 8700, Clarus 500.
Specifications
Specifications
| Max. Temperature | 350°C |
| Temperature Range | -60°C to +350°C |
| Length (Metric) | 2000 mm |
| Material | Siltek-treated Stainless Steel |
| Packing Material | 10% Rtx™-1 |
| Mesh Size | 100/120 Silcoport W |
| Diameter (Metric) Inner | 2 mm |
| Diameter (Metric) Outer | 3.17 mm |
| USP Type | G1, G2, G38 |
| Type | SRI 8610C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
