Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bonded Packed Column Stationary Phases: 10% Rtx-Stabilwax
SDP

Catalog No. 06714495 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packing Material:
10% Rtx™-Stabilwax
Silcoport™ W
Material:
SilcoSmooth™
Siltek-treated Stainless Steel
Stainless Steel
Mesh Size:
100/120
100/120 Silcoport W
For Use With (Equipment):
Agilent 5880, 5890, 5987, 6890, 7890
General
PE/Sigma
SRI 8610C
Shimadzu 14A, 2014
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packing Material Material Mesh Size For Use With (Equipment)
06-714-495 Silcoport™ W SilcoSmooth™ 100/120 PE/Sigma
06-714-602 Silcoport™ W Stainless Steel 100/120 General
06-714-493 10% Rtx™-Stabilwax SilcoSmooth™ 100/120 Silcoport W Agilent 5880, 5890, 5987, 6890, 7890
06-714-497 10% Rtx™-Stabilwax Siltek-treated Stainless Steel 100/120 Silcoport W Shimadzu 14A, 2014
06-714-498 10% Rtx™-Stabilwax Siltek-treated Stainless Steel 100/120 Silcoport W SRI 8610C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-714-495 Supplier Restek™ Supplier No. 80416830

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 10% Rtx-Stabilwax are for on-column injections.

  • Low bleed levels
  • Longer column lifetimes
  • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
  • Bonded packings usually stabilize in 30 minutes or less
  • Detectors can be operated at higher sensitivities due to low-bleed
  • Reduces need for data system bleed compensation
  • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
  • No sagging due to thorough cross-linking of material
  • Mesh: 100/120 Silcoport W

Order Info

Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

Column Configuration Key: General - General Configuration; Agilent - Agilent 5880, 5890, 5987, 6890, 7890; PE Auto Sys - PE Auto System 8300, 8400, 8700, Clarus 500.

Specifications

Max. Temperature 240°C
Temperature Range 40°C to 240°C
Length (Metric) 2 m
Material SilcoSmooth™
Packing Material Silcoport™ W
Mesh Size 100/120
Diameter (Metric) Inner 2 mm
Diameter (Metric) Outer 3.17 mm
Particle Size 2 μm
Stationary Phase 10% Rtx™-Stabilwax
For Use With (Equipment) PE/Sigma
Film Thickness 2 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.