Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bonded Packed Column Stationary Phases: 3% Rtx-1
SDP

Catalog No. 06714426 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packing Material:
3% Rtx™-1
Silcoport™ W
Material:
SilcoSmooth™
Siltek-treated Stainless Steel
Stainless Steel
Stationary Phase:
3% Rtx™-1
3% Rtx™-Stabilwax
Bonded Phase 3% Rtx™-1
For Use With (Equipment):
Agilent
Agilent 5880, 5890, 5987, 6890, 7890
General
PE/Sigma
SRI 9300
Varian
Varian 3700, Vista Series, FID
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packing Material Material Stationary Phase For Use With (Equipment)
06-714-426 3% Rtx™-1 SilcoSmooth™ Bonded Phase 3% Rtx™-1 Varian 3700, Vista Series, FID
06-714-422 Silcoport™ W SilcoSmooth™ 3% Rtx™-1 General
06-714-424 3% Rtx™-1 SilcoSmooth™ Bonded Phase 3% Rtx™-1 Agilent 5880, 5890, 5987, 6890, 7890
06-714-425 Silcoport™ W SilcoSmooth™ 3% Rtx™-1 Varian
06-714-427 Silcoport™ W SilcoSmooth™ 3% Rtx™-1 PE/Sigma
06-714-430 3% Rtx™-1 Siltek-treated Stainless Steel Bonded Phase 3% Rtx™-1 SRI 9300
06-714-587 Silcoport™ W Stainless Steel 3% Rtx™-Stabilwax Agilent
06-714-588 3% Rtx™-1 Stainless Steel Bonded Phase 3% Rtx™-1 Varian 3700, Vista Series, FID
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-714-426 Supplier Restek™ Supplier No. 80401820901

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 3% Rtx-1 are for on-column injections.

  • Low bleed levels
  • Longer column lifetimes
  • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
  • Bonded packings usually stabilize in 30 minutes or less
  • Detectors can be operated at higher sensitivities due to low-bleed
  • Reduces need for data system bleed compensation
  • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
  • No sagging due to thorough cross-linking of material
  • Mesh: 100/120 Silcoport W

Order Info

Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

Column Configuration Key: General - General Configuration; Agilent - Agilent 5880, 5890, 5987, 6890, 7890

Specifications

Max. Temperature 350°C
Temperature Range -60°C to +350°C
Length (Metric) 2000 mm
Material SilcoSmooth™
Packing Material 3% Rtx™-1
Mesh Size 100/120 Silcoport W
Diameter (Metric) Inner 2 mm
Diameter (Metric) Outer 3.17 mm
USP Type G1, G2, G38
Stationary Phase Bonded Phase 3% Rtx™-1
For Use With (Equipment) Varian 3700, Vista Series, FID
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.