Learn More
Restek™ Capillary FID Jets for Agilent™ 5890/6890/6850 GC
SDP
Catalog No. 06710892 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
High-Performance Siltek Treated
Standard
Packaging Quantity:
3 Pack
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
06-710-892 Standard 3 Pack
06-710-891 Standard Each
06-710-894 High-Performance Siltek Treated 3 Pack
06-710-893 High-Performance Siltek Treated Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-710-892 Supplier Restek™ Supplier No. 20671
Add to Cart
Add to Cart
Fluted tip to guide the capillary column into the jet
  • Threads specially coated for easy installation and removal
  • Standard version undergoes special processing to ensure the highest degree of cleanliness
  • High-performance version is Siltek™ treated to render extremely inert for use with active compounds
Specifications
Type Standard
Packaging Quantity 3 Pack
For Use With Agilent 5890/6890/6850