Learn More
Restek™ Capillary Installation Gauge for Agilent GCs
SDP
Catalog No. 06711376 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With:
Agilent GC
Varian GCs, 0.0625 in. Ferrules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
06-711-376 Agilent GC
06-729-309 Varian GCs, 0.0625 in. Ferrules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-711-376 Supplier Restek™ Supplier No. 21034
Seats graphite ferrules onto column for consistent installations.
  • Made from high-quality stainless steel
  • Seats graphite ferrules onto column for consistent installations
  • Prevents crushed column ends
  • Agilent-style fittings (compact ferrules)
Specifications
Type Gauge
For Use With Agilent GC