Learn More
Restek™ Corporation Vu2 Union™ Connector Kit Connectors
SDP
Catalog No. 06711920 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Inner:
0.15 to 0.25 mm
0.28 to 0.32 mm
0.45 to 0.5 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
06-711-920 0.15 to 0.25 mm
06-711-921 0.28 to 0.32 mm
06-711-922 0.45 to 0.5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-711-920 Supplier Restek™ Supplier No. 21105
Add to Cart
Add to Cart
Vu2 Union connector combines the simplicity of a Press-Tight union with the strength of a metal union.
  • Connect a guard column to an analytical column
  • Connect a column to a transfer line
  • Connect two columns in series
  • Repair a broken column
  • Fit both Restek cage designs
Specifications
Type Connector
Diameter (Metric) Inner 0.15 to 0.25 mm
Includes Vu2 Union body, two knurled nuts, two Press-Tight unions, and four ferrules
For Use With Vu2 Union Connector Kit