Description
Model 108
- Offers both 10.0 and 10.6 eV potential
- Base Dia.: 0.781 in.
- Used in Tracor, OI, and Baseline instruments
Model 103C
- Base Dia.: 1.375 in.
- Used in HNU and SRI detectors
Specifications
Specifications
| Type | Lamp |
| Diameter (English) Inner | 0.781 in. |
| Model | 108 |