Description
- High accuracy of dispensed volumes
- Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
Specifications
Specifications
| For Use With (Application) | For gases or liquids, for maximum inertness |
| Product Type | SGE PTFE Tip, Gas-Tight Syringes |
| Model | 100F-LL-GT |
| Volume (Metric) | 100 μL |
| Connection Type | Luer Lock |