Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ High-Pressure, Cup-Type In-Line Filters
SDP

Catalog No. 06712590 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Packaging Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
06-712-590 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-712-590 Supplier Restek™ Supplier No. 25000
Add to Cart
Add to Cart

Used in fluid streams operating to 15,000psi (103,421 kPa).

  • Large (2.5 sq. cm) surface area to give long operating lifetime
  • Mounted in screw-type adapters, they are easily removed for cleaning
  • Backflushing and cleaning in an ultrasonic bath with an appropriate solvent will restore them
  • Housings and all wetted parts are 316 stainless steel
  • Filters are packaged with gland nuts and ferrules
  • Bulkhead type fitting is available for thru-panel mounting
  • Tubing O.D.: 0.0625 in.
  • Connectors-CPI

Specifications

Type Filter
Diameter (English) Outer 1/16 in.
Porosity 0.5 μm
Packaging Quantity Each
Material Stainless Steel (316)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.