Learn More
Restek™ Inlet Liners for Splitless Injection—Siltek™ Gooseneck Splitless
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
Supplier: Restek™ 20799214
Description
- Compatible with Agilent™/Finnigan columns
- Liners should be packed with wool or fused silica beads to trap nonvolatile residue and prevent column contamination when analyzing dirty samples
- Include a Siltek—deactivated surface offer high temperature stability, extreme durability, and low bleed for minimized breakdown and better sample recovery
Specifications
Siltek Single Gooseneck Splitless Inlet Liner | |
5/Pk. | |
0.65 cm | |
Borosilicate Glass |
7.85 cm | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |