Learn More
Restek™ Jumbo Syringes
SDP
Catalog No. 06711501 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
06-711-501 500 mL
06-711-503 2000 mL
06-711-502 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-711-501 Supplier Restek™ Supplier No. 21275
Add to Cart
Add to Cart
Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.
  • Adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time
  • Central port supplied with a luer lock fitting
  • Secondary port is supplied with a septum nut, which enables access to gas sample for adding standards or removing a subsample
  • Detachable plunger stem for easy sample storage
Specifications
Product Type Syringe
Volume (Metric) 500 mL