Learn More
Restek™ Parker Balston™ Zero Air Generators: Replacement Catalyst Towers
SDP
Catalog No. 06711810 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Flow Rate:
1000 cc/min.
18000 cc/min.
30000 cc/min.
3500 cc/min.
7000 cc/min.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
06-711-810 1000 cc/min.
06-711-809 3500 cc/min.
06-711-811 7000 cc/min.
06-711-812 18000 cc/min.
06-711-813 30000 cc/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 06-711-810 Supplier Restek™ Supplier No. 22005
Add to Cart
Add to Cart
Replacement Catalyst Towers for Parker Balston Zero Air generators.
  • Flow Capacity Range: 1000 to 30,000 cc/min.
Specifications
Certifications/Compliance CE
Flow Rate 1000 cc/min.
For Use With (Equipment) 75-83na Zero Air Generator
Type Replacement Catalyst Tower