Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ PEEK™ Tubing Elbows
SDP

Catalog No. p-2386691
Click to view available options
Type:
Angle Tubing Elbow, 180°
Angle Tubing Elbow, 90°

Ideal for routing 1/16 in. PEEK tubing through your system.

  • Simply snap the tubing into the elbow
  • Prevent pinching of PEEK tubing, which can cause high pressure
  • Quantity: 5/Pk.

Specifications

Type Angle Tubing Elbow, 90°
Diameter (English) Inner 0.0625 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.