Learn More
Restek™ Presents Hamilton PTFE Fluorocarbon Resin-Tip, Gas-Tight Syringes: Needle Sold Separately
Shop All Restek Corporation Products
Description
- High accuracy of dispensed volumes
- Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
- Without slots
Specifications
Specifications
| Material | PTFE (Tip) |
| For Use With (Application) | Suitable for gases or liquids for maximum inertness |
| Product Type | Hamilton PTFE Tip, Gas-Tight Syringes |
| Model | 1005 |
| Quantity | Ea. |
| Volume (Metric) | 5 mL |
| Needle Point Style | No Slots |
| Connection Type | Luer Lock |