Learn More
Restek™ Presents SGE PTFE Fluorocarbon Resin-Tipped, Gas-Tight Syringe Replacement Needles
Shop All Restek Corporation Products

Description
- PTFE-tipped
- Stainless steel needle
- Nickel-plated brass hub
- Needle Point Style: 2
- Needle Length: 50mm
Specifications
Specifications
| Material | Nickel-plated Brass (Hub), Stainless Steel |
| Needle Gauge | 25 G |
| Length (English) Needle | 2 in. |
| Product Type | Replacement Syringe Needle |
| Volume (Metric) | 25 to 500 μL |
| Needle Point Style | Bevel |
| For Use With (Equipment) | Removable Needle Syringe |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.