Learn More
SGE PTFE Fluorocarbon Resin-Tipped, Gas-Tight Syringes from Restek™
Shop All Restek Corporation Products
Description
Suitable for gases or liquids, for maximum inertness.
- High accuracy of dispensed volumes
- Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
Specifications
Specifications
| Material | PTFE (Tip) |
| For Use With (Application) | Suitable for gases or liquids and high accuracy of dispensed volumes |
| Product Type | SGE PTFE Tip, Gas-Tight Syringes |
| Model | 1MDF-LL-GT |
| Quantity | Ea. |
| Volume (Metric) | 1 mL |
| Connection Type | Luer Lock |