Learn More
Restek™ Purge-and-Trap Spargers for Tekmar 2000, 3000 or 3100
SDP
Catalog No. 06611192 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Fritted
Non-Fritted
Capacity:
5 mL
10 mL
25 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Capacity
06-611-192 Non-Fritted 10 mL
06-711-453 Fritted 5 mL
06-711-454 Fritted 25 mL
06-611-191 Non-Fritted 5 mL
06-611-190 Fritted 10 mL
06-611-193 Non-Fritted 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-611-192 Supplier Restek™ Supplier No. 26140
Add to Cart
Add to Cart
For use with Tekmar purge-and-trap instruments only.
  • Available with uniform frits to ensure maximum purging efficiency
  • Use non-fritted spargers for wastewater samples
  • Manufactured to tight tolerances to ensure a leak-tight seal
  • Designed for use with Tekmar Purge-and-Trap instruments only
Specifications
Type Non-Fritted
Material Mixed
Mounting 1/2 in. mount
Capacity 10 mL
Quantity 1/Pk.
For Use With (Equipment) Tekmar Purge-and-Trap Instruments