Description
- Available with uniform frits to ensure maximum purging efficiency
- Use non-fritted spargers for wastewater samples
- Manufactured to tight tolerances to ensure a leak-tight seal
- Designed for use with Tekmar Purge-and-Trap instruments only
Specifications
Specifications
| Type | Non-Fritted |
| Material | Mixed |
| Mounting | 1/2 in. mount |
| Capacity | 25 mL |
| Quantity | 1/Pk. |
| For Use With (Equipment) | Tekmar Purge-and-Trap Instruments |