Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Shop All Restek Corporation Products
Click to view available options
Type:
O-rings for Trap Fittings
Replacement Split Vent Trap
Replacement Trap
Split Vent Trap Assembly
Description
- Built-in frits and adsorbents remove both moisture and hydrocarbons
- Trap can be regenerated to remove contaminants and restore performance
- Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Specifications
Type | Replacement Split Vent Trap |
For Use With (Application) | Gas Chromatography |
For Use With (Equipment) | Agilent 6890/6850 GCs (replacement for G1544-80550) |