Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
SDP
Catalog No. 06712138 Shop All Restek Corporation Products
Click to view available options
Type:
O-rings for Trap Fittings
Replacement Split Vent Trap
Replacement Trap
Split Vent Trap Assembly
Catalog No. 06-712-138
Supplier: Restek™ 23031
Only null left
Add to Cart
Add to Cart
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
  • Built-in frits and adsorbents remove both moisture and hydrocarbons
  • Trap can be regenerated to remove contaminants and restore performance
  • Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Type Split Vent Trap Assembly
For Use With (Application) Gas Chromatography
For Use With (Equipment) Agilent 6850/6890 GCs
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.