Description
- Increase column lifetime because oxygen cannot permeate into the carrier gas
- Reduced noise benefits high-sensitivity detectors (ECDs or MSDs)
- Special grade of stainless steel that is softer and deforms more easily, creating a better seal
- Gold-plated seals offer better inertness than standard stainless steel
- Siltek™ seals feature inertness similar to that of fused silica capillary columns
- All seals include washers
- 1.2mm inlet seal is recommended for use with Vespel™/graphite ferrules or when installing two columns using a two-hole ferrule
- 0.8mm inlet seal is recommended for use with graphite ferrules and single capillary column installation
Specifications
Specifications
| Type | Seal |
| Diameter (Metric) Inner | 0.8 mm |
| Packaging Quantity | 10/Pk. |
| Material | Gold-plated |