Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ RTX™-20 Capillary Columns
SDP

Catalog No. 06708786 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Length (Metric):
20 m
30 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Particle Size:
0.25 μm
1 μm
3 μm
Includes:
5 m Guard Column with Press-Tight, Glue
5m Integra guard column
Not Applicable
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Inner Particle Size Includes
06-708-786 20 m 0.32 mm 0.25 μm Not Applicable
06-708-783 30 m 0.25 mm 0.25 μm 5m Integra guard column
06-708-812 30 m 0.32 mm 1 μm 5m Integra guard column
06-708-810 30 m 0.25 mm 1 μm Not Applicable
06-708-834 30 m 0.53 mm 3 μm 5m Integra guard column
06-708-813 30 m 0.32 mm 1 μm 5 m Guard Column with Press-Tight, Glue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-708-786 Supplier Restek™ Supplier No. 10324112

Tight mono-modal distribution and extremely low bleed

  • Low to midpolarity phase; Crossbond™ diphenyl dimethyl polysiloxane
  • General-purpose columns for volatile compounds, flavor compounds, alcoholic beverages
  • All residual catalysts and low molecular weight fragments are removed from the polymer
  • Temperature range: -20°C to 320°C (-4°F to 608°F)
  • Equivalent to USP G28, G32 phases
  • Fused silica

Specifications

For Use With (Application) Crossbond™, Flavor Compounds Analysis, Alcoholic Beverages Analysis
Particle Size 0.25 μm
Film Thickness 0.25 μm
Length (Metric) 20 m
Material Stainless Steel
Packing Material Rtx™-20
Polarity Low to mid polar
Coating Fused Silica
Includes Not Applicable
USP Type G28, G32
Stationary Phase 20% Diphenyl/80% Dimethyl Polysiloxane
Diameter (Metric) Inner 0.32 mm
Max. Temperature (Metric) 350°C
Temperature Range (Metric) -20°C to +300/320°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.