Learn More
Description
- Low to midpolarity phase; Crossbond™ diphenyl dimethyl polysiloxane
- General-purpose columns for volatile compounds, flavor compounds, alcoholic beverages
- All residual catalysts and low molecular weight fragments are removed from the polymer
- Temperature range: -20°C to 320°C (-4°F to 608°F)
- Equivalent to USP G28, G32 phases
- Fused silica
Specifications
Specifications
| For Use With (Application) | Crossbond™, Flavor Compounds Analysis, Alcoholic Beverages Analysis |
| Particle Size | 1 μm |
| Film Thickness | 1 μm |
| Length (Metric) | 30 m |
| Material | Stainless Steel |
| Packing Material | Rtx™-20 |
| Polarity | Low to mid polar |
| Coating | Fused Silica |
| Includes | 5m Integra guard column |
| USP Type | G28, G32 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
