Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ RTX™-5 Capillary Columns, 30 m
SDP

Catalog No. 06708757 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
1 μm
1.5 μm
3 μm
5 μm
Stationary Phase:
5% Diphenyl/95% Dimethyl Polysiloxane
Rtx™-5
Includes:
10m Integra-Guard Column
5m Integra-Guard Column
Cage
Column Only
Integra Guard Column
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Film Thickness Stationary Phase Includes
06-708-757 0.53 mm 3 μm 5% Diphenyl/95% Dimethyl Polysiloxane Integra Guard Column
06-708-672 0.25 mm 1 μm Rtx™-5 5m Integra-Guard Column
06-708-673 0.25 mm 1 μm Rtx™-5 10m Integra-Guard Column
06-708-679 0.32 mm 1 μm Rtx™-5 5m Integra-Guard Column
06-708-691 0.53 mm 1 μm Rtx™-5 10m Integra-Guard Column
06-708-719 0.53 mm 1.5 μm Rtx™-5 10m Integra-Guard Column
06-708-735 0.53 mm 5 μm Rtx™-5 10m Integra-Guard Column
06-708-737 0.53 mm 5 μm Rtx™-5 Column Only
06-717-561 0.53 mm 1 μm Rtx™-5 Column Only
06-708-680 0.32 mm 1 μm 5% Diphenyl/95% Dimethyl Polysiloxane Integra Guard Column
06-708-714 0.32 mm 1.5 μm 5% Diphenyl/95% Dimethyl Polysiloxane Integra Guard Column
06-708-715 0.32 mm 1.5 μm 5% Diphenyl/95% Dimethyl Polysiloxane Integra Guard Column
06-708-716 0.32 mm 1.5 μm Rtx™-5 Cage
06-717-559 0.25 mm 1 μm 5% Diphenyl/95% Dimethyl Polysiloxane Column Only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 06-708-757 Supplier Restek™ Supplier No. 10285129

General-purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, semivolatiles

  • Low polarity phase; Crossbond™ diphenyl dimethyl polysiloxane)
  • General-purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g., Aroclor mixes), essential oils, semivolatiles
  • Diphenyl dimethyl polysiloxane stationary phase is the most popular GC stationary phase and is used in a wide variety of applications
  • All residual catalysts and low molecular weight fragments are removed from the Rtx™-5 polymer, providing a tight mono-modal distribution and extremely low bleed
  • Temperature range: -60°C to 350°C (-76°F to 662°F)
  • Equivalent to USP G27 and G36 phases

Specifications

For Use With (Application) Crossbond™, Solvent Impurities Analysis, Pesticide Analysis, Hydrocarbon Analysis, PCB Congener Analysis, Essential Oils, Semivolatiles Analysis
Film Thickness 3 μm
Length (Metric) 30 m
Material Stainless Steel
Packing Material Rtx™-5
Polarity Low polar
Coating Fused Silica
Includes Integra Guard Column
USP Type G27, G36
Stationary Phase 5% Diphenyl/95% Dimethyl Polysiloxane
Diameter (Metric) Inner 0.53 mm
Max. Temperature (Metric) 350°C
Temperature Range (Metric) -60°C to +270/290°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.